Gold ≥99.99% (trace metals basis), gauze woven from wire 100 mesh Ø 0.064 mm
Supplier: BeanTown Chemical
137620-25X25MM
137620-50X100
137620-50X50MM
BT137620-25X25EA
291
USD
BT137620-25X25
BT137620-50X10
BT137620-50X50
Gold ≥99.99% (trace metals basis), gauze woven from wire 100 mesh Ø 0.064 mm
Gold
CAS: 7440-57-5; EC No: 231-165-9; MDL No: MFCD00003436; RTECS: MD5420000
Wire Cloth, 100x100mm, ~4.9g; Molecular Formula: Au; MW: 196.97
Melting Point: 1063°; Boiling Point: 2808°
Density (g/mL): 19.3
- RTECS: MD5420000
- Wire Cloth, 100x100mm, ~4.9g
Formula:
Au Boiling Pt: 2808 °C Melting Pt: 1063 °C Density: 19.3 g/mL |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...