Zinc fluoride ≥99.995%
Supplier: BeanTown Chemical
137550-100G
137550-25G
137550-5G
BT137550-100GEA
590
USD
BT137550-100G
BT137550-25G
BT137550-5G
Zinc fluoride ≥99.995%
Zinc fluoride
CAS: 7783-49-5; EC No: 232-001-9; MDL No: MFCD00011298; RTECS: ZH3500000
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: ZnF2; MW: 103.39
Melting Point: 872°; Boiling Point: 1500°
Density (g/mL): 4.95
- RTECS: ZH3500000
- Powder
Formula:
F₂Zn Boiling Pt: 1500 °C Melting Pt: 872 °C Density: 4.95 g/mL |
MDL Number:
MFCD00011298 CAS Number: 7783-49-5 EINECS: 232-001-9 UN: 3288 ADR: 6.1,III |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...