Ferric hypophosphite

Supplier: BeanTown Chemical

Synonyms: Iron(III) hypophosphite

137545-100G
BT137545-100GEA 272 USD
BT137545-100G BT137545-25G
Ferric hypophosphite
Ferric hypophosphite

CAS: 7783-84-8; EC No: 232-030-7; MDL No: MFCD00049454 Powder; Linear Formula: H6FeO6P3; MW: 250.81


  • Powder

Formula: FeO₆P₃
CAS Number: 7783-84-8
EINECS: 232-030-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR