Diisopropyl ether ≥99%

Supplier: BeanTown Chemical

Synonyms: Isopropyl ether

137495-1L
BT137495-1LEA 154.74 USD
BT137495-1L
Diisopropyl ether ≥99%
Diisopropyl ether

CAS: 108-20-3; EC No: 203-560-6; MDL No: MFCD00008880; RTECS: TZ5425000 UN No: UN1159; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)2CHOCH(CH3)2; Molecular Formula: C6H14O; MW: 102.17 Melting Point: -85°; Boiling Point: 68-69°; Flash point: -29°C (-20°F) Density (g/mL): 0.725; Refractive Index: 1.367


  • RTECS: TZ5425000
  • Refractive index: 1.367
  • Liquid

Formula: (CH₃)₂CHOCH(CH₃)₂
MW: 102.18 g/mol
Boiling Pt: 68.5
Melting Pt: -86
Density: 0.725 g/mL
Flash Pt: –29 °C (–20 °F)
MDL Number: MFCD00008880
CAS Number: 108-20-3
EINECS: 203-560-6
UN: 1159
ADR: 3 II
Merck Index: 13,05232
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=932b8c021b2bfa9a11b8d8812cafbafecc6f9239&vid=dea267e876203eb2c2674787701ec602da1b3cf6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR