Samarium ≥99.9% (trace rare earth metals basis), pieces, sublimed dendritic

Supplier: BeanTown Chemical

137480-10G 137480-250G 137480-50G
BT137480-10GEA 56.06 USD
BT137480-10G BT137480-250G BT137480-50G
Samarium ≥99.9% (trace rare earth metals basis), pieces, sublimed dendritic
Samarium

CAS: 7440-19-9; EC No: 231-128-7; MDL No: MFCD00011233 UN No: UN3178; Haz Class: 4.1; Packing Group: II Solid; Molecular Formula: Sm; MW: 150.36 Melting Point: 1074°; Boiling Point: 1794° Density (g/mL): 7.47


  • Solid

Formula: Sm
MW: 150.36 g/mol
Boiling Pt: 1900
Melting Pt: 1076.85
Density: 7.47 g/mL
MDL Number: MFCD00011233
CAS Number: 7440-19-9
EINECS: 231-128-7
ADR: 4.1 II
Merck Index: 12,08498

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR