1-Bromopyrene ≥95%
Supplier: BeanTown Chemical
137430-1G
137430-5G
BT137430-1GEA
48
USD
BT137430-1G
BT137430-5G
1-Bromopyrene ≥95%
1-Bromopyrene
CAS: 1714-29-0; MDL No: MFCD00015767
Crystalline; Molecular Formula: C16H9Br; MW: 281.15
Melting Point: 102-105°
- Crystalline
Formula:
C₁₆H₉Br Melting Pt: 102…105 °C |
MDL Number:
MFCD00015767 CAS Number: 1714-29-0 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...