Gallium ≥99.99% (trace metals basis), ingot

Supplier: BeanTown Chemical

137400-250G 137400-25G 137400-50G
BT137400-250GEA 789.84 USD
BT137400-250G BT137400-25G BT137400-50G
Gallium ≥99.99% (trace metals basis), ingot
Gallium
Formula: Ga
Boiling Pt: 2403 °C
Melting Pt: 29.8 °C
Density: 5.904 g/mL
MDL Number: MFCD00134045
CAS Number: 7440-55-3
EINECS: 231-163-8
UN: 2803
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR