4-Cyano-3,5-difluorophenyl-4-n-pentylbenzoate ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-Cyano-3,5-difluorophenyl 4-Amylbenzoate, 4-Cyano-3,5-difluorophenyl 4-Pentylbenzoate, 4-Amylbenzoic acid 4-Cyano-3,5-difluorophenyl ester, 4-Pentylbenzoic acid 4-Cyano-3,5-difluorophenyl ester
137395-1G
137395-5G
BT137395-1GEA
103
USD
BT137395-1G
BT137395-5G
4-Cyano-3,5-difluorophenyl-4-n-pentylbenzoate ≥97%
4-Cyano-3,5-difluorophenyl-4-n-pentylbenzoate
CAS: 123843-69-6; MDL No: MFCD11053375
Solid; Molecular Formula: C19H17F2NO2; MW: 329.35
Melting Point: 45°
- Solid
Formula:
C₁₉H₁₇F₂NO₂ Melting Pt: 45 °C |
MDL Number:
MFCD11053375 CAS Number: 123843-69-6 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...