(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) ≥97%
Supplier: BeanTown Chemical
Synonyms:
XANTPHOS, 9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene
137355-1G
137355-5G
BT137355-1GEA
66
USD
BT137355-1G
BT137355-5G
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) ≥97%
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine)
CAS: 161265-03-8; MDL No: MFCD00233866
Crystalline; Molecular Formula: C39H32OP2; MW: 578.62
Melting Point: 224-228°
- Crystalline
Formula:
C₃₉H₃₂OP₂ MW: 578.62 g/mol Melting Pt: 224…228 °C |
MDL Number:
MFCD00233866 CAS Number: 161265-03-8 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...