2-Decanone ≥97%
Supplier: BeanTown Chemical
Synonyms:
Decan-2-one
137290-100G
137290-25G
137290-5G
BT137290-100GEA
193
USD
BT137290-100G
BT137290-25G
BT137290-5G
2-Decanone ≥97%
2-Decanone
CAS: 693-54-9; EC No: 211-752-6; MDL No: MFCD00009571; RTECS: HE0725000
Liquid; Linear Formula: CH3(CH2)7COCH3; Molecular Formula: C10H20O; MW: 156.27
Melting Point: 3°; Boiling Point: 211°; Flash point: 86°C (187°F)
Density (g/mL): 0.825; Refractive Index: 1.425
- RTECS: HE0725000
- Refractive index: 1.425
- Liquid
Formula:
C₁₀H₂₀O Boiling Pt: 211 °C Melting Pt: 3 °C Density: 0.825 g/mL Flash Pt: 86 °C (187 °F) |
MDL Number:
MFCD00009571 CAS Number: 693-54-9 EINECS: 211-752-6 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...