Formamide ≥99%

Supplier: BeanTown Chemical

Synonyms: Carbamaldehyde

137285-10KG 137285-2.5KG 137285-500G
BT137285-10KGEA 257 USD
BT137285-10KG BT137285-2.5KG BT137285-500G
Formamide ≥99%
Formamide

CAS: 75-12-7; EC No: 200-842-0; MDL No: MFCD00007941; RTECS: LQ0525000 Liquid; Linear Formula: HCONH2; MW: 45.04 Melting Point: 2-3°; Boiling Point: 210°; Flash point: 175°C (347°F) Density (g/mL): 1.134; Refractive Index: 1.447 Hygroscopic


  • RTECS: LQ0525000
  • Refractive index: 1.447
  • Hygroscopic
  • Liquid

Formula: HCONH₂
MW: 45.04 g/mol
Boiling Pt: 210
Melting Pt: 2
Density: 1.134 g/mL
Flash Pt: 175 °C (347 °F)
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR