3-Noradamantanecarboxylic acid ≥98%
Supplier: BeanTown Chemical
137275-1G
137275-250MG
137275-5G
BT137275-1GEA
91
USD
BT137275-1G
BT137275-250MG
BT137275-5G
3-Noradamantanecarboxylic acid ≥98%
3-Noradamantanecarboxylic acid
CAS: 16200-53-6; MDL No: MFCD00213463
Solid; Molecular Formula: C10H14O2; MW: 166.22
Melting Point: 105-107°
- Solid
Formula:
C₁₀H₁₄O₂ Melting Pt: 105…107 °C |
MDL Number:
MFCD00213463 CAS Number: 16200-53-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...