Boron trifluoride diethyl ether complex ≥98%
Supplier: BeanTown Chemical
|
|
Synonyms:
Boron fluoride, ethyl ether complex, Boron trifluoride ethyl etherate, Boron trifluoride diethyl ether, Boron trifluoride etherate, Diethyl ether boron trifluoride, Boron trifluoride diethyl etherate
137235-100ML
BT137235-100MLEA
37.75
USD
BT137235-100ML
BT137235-500ML
Boron trifluoride diethyl ether complex ≥98%
Boron trifluoride diethyl ether complex
CAS: 109-63-7; EC No: 203-689-8; MDL No: MFCD00013194
UN No: UN2604; Haz Class: 8 (3); Packing Group: I
Liquid; Linear Formula: BF3·O(CH2CH3)2; Molecular Formula: C4H10BF3O; MW: 141.93
Melting Point: -58°; Boiling Point: 126-129°; Flash point: 48°C (118°F)
Density (g/mL): 1.15; Refractive Index: 1.344
Moisture Sensitive
- Refractive index: 1.344
- Moisture Sensitive
- Liquid
Formula:
C₄H₁₀BF₃O Boiling Pt: 126…129 °C Melting Pt: –58 °C Density: 1.15 g/mL Flash Pt: 48 °C (118 °F) |
MDL Number:
MFCD00013194 CAS Number: 109-63-7 EINECS: 203-689-8 UN: 2604 ADR: 8,I |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...