1H,1H,2H,2H-Perfluorooctyltriethoxysilane ≥97%
Supplier: BeanTown Chemical
Synonyms:
Triethoxy-1H,1H,2H,2H-tridecafluoro-n-octylsilane, Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-triethoxysilane, Triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane;
137180-25G
137180-5G
BT137180-25GEA
309.95
USD
BT137180-25G
BT137180-5G
1H,1H,2H,2H-Perfluorooctyltriethoxysilane ≥97%
1H,1H,2H,2H-Perfluorooctyltriethoxysilane
CAS: 51851-37-7; EC No: 257-473-3; MDL No: MFCD00042333
Liquid; Molecular Formula: C14H19F13O3Si; MW: 510.36
Flash point: 97°C (207°F)
Density (g/mL): 1.33; Refractive Index: 1.344
Moisture Sensitive
- Refractive index: 1.344
- Moisture Sensitive
- Liquid
Formula:
C₁₄H₁₉F₁₃O₃Si MW: 510.367 g/mol Density: 1.33 g/mL Flash Pt: 97 °C (207 °F) |
MDL Number:
MFCD00042333 CAS Number: 51851-37-7 EINECS: 257-473-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...