(S)-2-Amino-6-guanidinohexanoic acid hydrochloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
H-HoArg-OH·HCl, L(+)-Homoarginine hydrochloride, H-HomoArg-OH.HCl, H-HomoArg-OH·HCl
137160-1G
137160-25G
137160-5G
BT137160-1GEA
65
USD
BT137160-1G
BT137160-25G
BT137160-5G
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride ≥98%
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride
CAS: 1483-01-8; EC No: 216-045-6; MDL No: MFCD00012622
Powder; Molecular Formula: C7H17ClN4O2; MW: 224.69
Melting Point: 213-215°
- Powder
Formula:
C₇H₁₆N₄O₂·HCl MW: 224.7 g/mol Melting Pt: 213…215 °C |
MDL Number:
MFCD00012622 CAS Number: 1483-01-8 EINECS: 216-045-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...