Diammonium hydrogen orthophosphate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate
137150-500G
BT137150-500GEA
39.97
USD
BT137150-500G
Diammonium hydrogen orthophosphate ≥98%
di-Ammonium hydrogen orthophosphate
CAS: 7783-28-0; EC No: 231-987-8; MDL No: MFCD00010891
Crystalline; Linear Formula: (NH4)2HPO4 ; MW: 132.06
Melting Point: 155° (decomposes)
- Crystalline
Formula:
(NH₄)₂HPO₄ Melting Pt: 155 °C (dec.) |
MDL Number:
MFCD00010891 CAS Number: 7783-28-0 EINECS: 231-987-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...