Tris(triphenylphosphine)rhodium(I) chloride ≥97%
Supplier: BeanTown Chemical
137115-1G
137115-5G
BT137115-1GEA
148
USD
BT137115-1G
BT137115-5G
Tris(triphenylphosphine)rhodium(I) chloride ≥97%
Tris(triphenylphosphine)rhodium (I) chloride
CAS: 14694-95-2; EC No: 238-744-5; MDL No: MFCD00010016
Crystalline; Linear Formula: [(C6H5)3P]3RhCl; Molecular Formula: C54H45ClP3Rh; MW: 925.23
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
C₅₄H₄₅ClP₃Rh |
MDL Number:
MFCD00010016 CAS Number: 14694-95-2 EINECS: 238-744-5 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...