Tris(triphenylphosphine)rhodium(I) chloride ≥97%

Supplier: BeanTown Chemical

137115-1G 137115-5G
BT137115-1GEA 148 USD
BT137115-1G BT137115-5G
Tris(triphenylphosphine)rhodium(I) chloride ≥97%
Tris(triphenylphosphine)rhodium (I) chloride

CAS: 14694-95-2; EC No: 238-744-5; MDL No: MFCD00010016 Crystalline; Linear Formula: [(C6H5)3P]3RhCl; Molecular Formula: C54H45ClP3Rh; MW: 925.23 Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: C₅₄H₄₅ClP₃Rh
MDL Number: MFCD00010016
CAS Number: 14694-95-2
EINECS: 238-744-5

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR