trans-4-(4-Propylcyclohexyl)benzonitrile ≥99%

Supplier: BeanTown Chemical

Synonyms: 4-(trans-4-propylcyclohexyl)benzonitrile

137065-1G 137065-5G
BT137065-1GEA 83 USD
BT137065-1G BT137065-5G
trans-4-(4-Propylcyclohexyl)benzonitrile ≥99%
trans-4-(4-Propylcyclohexyl)benzonitrile

CAS: 61203-99-4; EC No: 262-654-5; MDL No: MFCD04038799 Solid; Molecular Formula: C16H21N; MW: 227.35 Melting Point: 45°


  • Solid

Formula: C₁₆H₂₁N
MW: 227.3446 g/mol
Melting Pt: 45 °C
MDL Number: MFCD04038799
CAS Number: 61203-99-4
EINECS: 262-654-5

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR