Sodium octyl sulfate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Octyl sulphate, sodium salt, Octyl sulfate, sodium salt, Octyl sulfuric acid salt, Octyl sulphuric acid salt
137025-1G
137025-25G
137025-5G
BT137025-1GEA
29
USD
BT137025-1G
BT137025-25G
BT137025-5G
Sodium octyl sulfate ≥99%
Sodium octyl sulphate
CAS: 142-31-4; EC No: 205-535-5; MDL No: MFCD00007470; RTECS: WT1175000
Crystalline; Linear Formula: CH3(CH7)OSO3Na; Molecular Formula: C8H17NaO4S; MW: 232.28
Melting Point: 195° (decomposes)
- RTECS: WT1175000
- Crystalline
Formula:
C₈H₁₇NaO₄S Melting Pt: 195 °C (dec.) |
MDL Number:
MFCD00007470 CAS Number: 142-31-4 EINECS: 205-535-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...