2-Hydroxypyridine ≥98%
Supplier: BeanTown Chemical
Synonyms:
Pyridin-2-one, Pyridin-2(1H)-one, α-Hydroxypyridine, 2-Pyridone, α-Pyridone
137020-100G
137020-25G
137020-500G
BT137020-100GEA
112
USD
BT137020-100G
BT137020-25G
BT137020-500G
2-Hydroxypyridine ≥98%
2-Hydroxypyridine
CAS: 142-08-5; EC No: 205-520-3; MDL No: MFCD00006268; RTECS: UV1144050
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Crystalline/Powder; Molecular Formula: C5H5NO; MW: 95.10
Melting Point: 105-107°; Boiling Point: 280-281°
- RTECS: UV1144050
- Crystalline/Powder
Formula:
C₅H₅NO Boiling Pt: 280…281 °C Melting Pt: 105…107 °C |
MDL Number:
MFCD00006268 CAS Number: 142-08-5 EINECS: 205-520-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...