Sodium dichromate dihydrate 99.5-100.5% ACS
Supplier: BeanTown Chemical
136980-100G
136980-2KG
136980-500G
BT136980-100GEA
39
USD
BT136980-100G
BT136980-2KG
BT136980-500G
Sodium dichromate dihydrate 99.5-100.5% ACS
Sodium dichromate dihydrate
CAS: 7789-12-0; EC No: 234-190-3; MDL No: MFCD00149166; RTECS: HX7750000
UN No: UN3087; Haz Class: 5.1 (6.1); Packing Group: II
Crystalline; Linear Formula: Na2Cr2O7·2H2O; MW: 298.00
Melting Point: 91°
Density (g/mL): 2.350
Hygroscopic
- RTECS: HX7750000
- Hygroscopic
- Crystalline
Formula:
Na₂Cr₂O₇·2H₂O Melting Pt: 91 °C Density: 2.350 g/mL |
MDL Number:
MFCD00149166 CAS Number: 7789-12-0 EINECS: 234-190-3 UN: 3087 ADR: 5.1,II |
Specification Test Results
Assay | 99.5-100.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...