Tetrabutylammonium dihydrogen phosphate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate
136970-10G
136970-2G
136970-500MG
BT136970-10GEA
270.33
USD
BT136970-10G
BT136970-2G
BT136970-500MG
Tetrabutylammonium dihydrogen phosphate ≥99%
Tetrabutylammonium dihydrogen phosphate
CAS: 5574-97-0; EC No: 226-947-1; MDL No: MFCD00064526
Crystalline; Linear Formula: [CH3(CH2)3]4N(H2PO4); Molecular Formula: C16H38NO4P; MW: 339.46
Melting Point: 151-154°
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
C₁₆H₃₈NO₄P MW: 339.46 g/mol Melting Pt: 151…154 °C |
MDL Number:
MFCD00064526 CAS Number: 5574-97-0 EINECS: 226-947-1 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...