Gold ≥99.99% (trace metals basis), foil, Thickness 0.127 mm

Supplier: BeanTown Chemical

136940-25X25MM 136940-50X50MM
BT136940-25X25EA 378 USD
BT136940-25X25 BT136940-50X50
Gold ≥99.99% (trace metals basis), foil, Thickness 0.127 mm
Gold

CAS: 7440-57-5; EC No: 231-165-9; MDL No: MFCD00003436; RTECS: MD5420000 Solid; Molecular Formula: Au; MW: 196.97 Melting Point: 1063°; Boiling Point: 2808° Density (g/mL): 19.3


  • RTECS: MD5420000
  • Solid

Formula: Au
Boiling Pt: 2808 °C
Melting Pt: 1063 °C
Density: 19.3 g/mL
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR