Gold ≥99.99% (trace metals basis), foil, Thickness 0.127 mm
Supplier: BeanTown Chemical
136940-25X25MM
136940-50X50MM
BT136940-25X25EA
378
USD
BT136940-25X25
BT136940-50X50
Gold ≥99.99% (trace metals basis), foil, Thickness 0.127 mm
Gold
CAS: 7440-57-5; EC No: 231-165-9; MDL No: MFCD00003436; RTECS: MD5420000
Solid; Molecular Formula: Au; MW: 196.97
Melting Point: 1063°; Boiling Point: 2808°
Density (g/mL): 19.3
- RTECS: MD5420000
- Solid
Formula:
Au Boiling Pt: 2808 °C Melting Pt: 1063 °C Density: 19.3 g/mL |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...