Barium titanate ≥99%

Supplier: BeanTown Chemical

Synonyms: Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide

136930-2KG 136930-500G 136930-50G
BT136930-2KGEA 136 USD
BT136930-2KG BT136930-500G BT136930-50G
Barium titanate ≥99%
Barium titanate

CAS: 12047-27-7; EC No: 234-975-0; MDL No: MFCD00003447 UN No: UN1564; Haz Class: 6.1; Packing Group: III -325 Mesh Powder ; Molecular Formula: BaTiO3; MW: 233.19 Density (g/mL): 6.08


  • -325 Mesh Powder

Formula: BaO₃Ti
Density: 6.08 g/mL
MDL Number: MFCD00003447
CAS Number: 12047-27-7
EINECS: 234-975-0
UN: 1564
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR