Pantoprazole sulfide
Supplier: BeanTown Chemical
Synonyms:
5-Difluoromethoxy-2-[(3,4-dimethoxy-2-pyridyl)methyl]thio-1H-benzimidazole, 5-(Difluoromethoxy)-2-(((3,4-dimethoxypyridin-2-yl)methyl)thio)-1H-benzo[d]imidazole, 5-(Difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridyl)methyl]thio]benzimidazole
136870-1G
136870-5G
BT136870-1GEA
25
USD
BT136870-1G
BT136870-5G
Pantoprazole sulfide
Pantoprazole sulfide
CAS: 102625-64-9; MDL No: MFCD07368273
Solid; Molecular Formula: C16H15F2N3O3S; MW: 367.37
Melting Point: 110-118°
- Solid
Formula:
C₁₆H₁₅F₂N₃O₃S MW: 367.37 g/mol Melting Pt: 110…118 °C |
MDL Number:
MFCD07368273 CAS Number: 102625-64-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...