(2-Naphthoxy)acetic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
2-Naphthoxyacetic acid
136785-100G
136785-2.5KG
136785-500G
BT136785-100GEA
41
USD
BT136785-100G
BT136785-2.5KG
BT136785-500G
(2-Naphthoxy)acetic acid ≥97%
(2-Naphthoxy)acetic acid
CAS: 120-23-0; EC No: 204-380-0; MDL No: MFCD00004066; RTECS: AI9659000
Crystalline; Molecular Formula: C12H10O3; MW: 202.21
Melting Point: 151-154°
- RTECS: AI9659000
- Crystalline
Formula:
C₁₂H₁₀O₃ Melting Pt: 151…154 °C |
MDL Number:
MFCD00004066 CAS Number: 120-23-0 EINECS: 204-380-0 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...