Tungsten trioxide ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Tungsten (VI) oxide
136700-100G
136700-2KG
136700-500G
BT136700-100GEA
62
USD
BT136700-100G
BT136700-2KG
BT136700-500G
Tungsten trioxide ≥99.5%
Tungsten trioxide
CAS: 1314-35-8; EC No: 215-231-4; MDL No: MFCD00011466; RTECS: YO7760000
10-20 Micron Powder; Linear Formula: WO3; MW: 231.85
Melting Point: 1472°; Boiling Point: 1837°
Density (g/mL): 7.16
Moisture Sensitive
- RTECS: YO7760000
- Moisture Sensitive
- 10-20 Micron Powder
Formula:
O₃W Boiling Pt: 1837 °C Melting Pt: 1472 °C Density: 7.16 g/mL |
MDL Number:
MFCD00011466 CAS Number: 1314-35-8 EINECS: 215-231-4 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...