EDTA disodium salt dihydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate

136615-1KG 136615-250G 136615-5KG
BT136615-1KGEA 116.95 USD
BT136615-1KG BT136615-250G BT136615-5KG
EDTA disodium salt dihydrate ≥99%
EDTA disodium salt dihydrate

CAS: 6381-92-6; EC No: 205-358-3; MDL No: MFCD00150037; RTECS: AH4375000 Powder; Molecular Formula: C10H14N2Na2O8·2H2O; MW: 372.24 Melting Point: 248° (decomposes)


  • RTECS: AH4375000
  • Powder

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Melting Pt: 255 (dec.)
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR