Lidocaine hydrochloride monohydrate

Supplier: BeanTown Chemical

136580-100G 136580-25G 136580-5G
BT136580-100GEA 235.42 USD
BT136580-100G BT136580-25G BT136580-5G
Lidocaine hydrochloride monohydrate
Lidocaine hydrochloride monohydrate

CAS: 6108-05-0; EC No: 200-803-8; MDL No: MFCD00150329; RTECS: AN7700000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline/Powder; Molecular Formula: C14H22N2O·HCl·H2O; MW: 288.82


  • RTECS: AN7700000
  • Crystalline/Powder

Formula: C₁₄H₂₃ClN₂O·1H₂O
MW: 270.8 g/mol
MDL Number: MFCD00150329
CAS Number: 6108-05-0
EINECS: 200-803-8
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR