Hafnium, Plasma Standard Solution, Hf 1,000 µg/ml

Supplier: BeanTown Chemical

136535-500ML
BT136535-100MLEA 109.26 USD
BT136535-100ML BT136535-500ML BT136535-50ML
Hafnium, Plasma Standard Solution, Hf 1,000 µg/ml
Standards Cationic Standards Hafnium Standards

EC No: 231-595-7
Haz class: 8
Packing group: III
GHS symbol: GHS05

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR