Tetrabutylammonium hydroxide 1.0 M in water ACS

Supplier: BeanTown Chemical

136520-100ML 136520-25ML 136520-500ML
BT136520-100MLEA 196.21 USD
BT136520-100ML BT136520-25ML BT136520-500ML
Tetrabutylammonium hydroxide 1.0 M in water ACS
Tetrabutylammonium hydroxide

CAS: 2052-49-5; EC No: 218-147-6; MDL No: MFCD00009425; RTECS: BS5425000 UN No: UN3267; Haz Class: 8; Packing Group: II Liquid; Linear Formula: [CH3(CH2)3]4N(OH); Molecular Formula: C16H37NO; MW: 259.48 Boiling Point: 100° Density (g/mL): 1; Refractive Index: 1.389 Air Sensitive


  • RTECS: BS5425000
  • Refractive index: 1.389
  • Air Sensitive
  • Liquid

Formula: C₁₆H₃₇NO
Boiling Pt: >100 °C
Density: 1 g/mL
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR