Tin(II) tartrate hydrate ≥95%
Supplier: BeanTown Chemical
136475-100G
136475-25G
BT136475-100GEA
384
USD
BT136475-100G
BT136475-25G
Tin(II) tartrate hydrate ≥95%
Tin(II) tartrate hydrate
CAS: 815-85-0; EC No: 212-427-1; MDL No: MFCD00054358; RTECS: EJ9892500
Powder; Molecular Formula: C4H4O6Sn·xH2O; MW: 266.78 (anhydrous)
Air Sensitive
- RTECS: EJ9892500
- Air Sensitive
- Powder
Formula:
C₄H₄O₆Sn·xH₂O |
MDL Number:
MFCD00054358 CAS Number: 815-85-0 EINECS: 212-427-1 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...