N,N-Dimethylformamide ≥99.8% ACS
Supplier: BeanTown Chemical
CAS: 68-12-2; EC No: 200-679-5; MDL No: MFCD00003284; RTECS: LQ2100000
UN No: UN2265; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: HCON(CH3)2 ; Molecular Formula: C3H7NO; MW: 73.09
Melting Point: -61°; Boiling Point: 153°; Flash point: 57°C (135°F)
Density (g/mL): 0.944; Refractive Index: 1.430
- RTECS: LQ2100000
- Refractive index: 1.430
- Liquid
Formula:
HCON(CH₃)₂ Boiling Pt: 153 °C Melting Pt: –61 °C Density: 0.944 g/mL Flash Pt: 57 °C (135 °F) |
MDL Number:
MFCD00003284 CAS Number: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3,III |
|
Specification Test Results
Assay | 99.8% min. |
Color (APHA) | ≤10 |
Residue after evaporation | ≤50ppm |
Titrable acid | ≤0.0005meq/g |
Titrable base | ≤0.003meq/g |
Specific gravity (20/20) | Per lot |
Methanol | Per lot |
Iron (Fe) | Per lot |
Water | ≤0.15% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...