Silica gel desiccant, with indicator

Supplier: BeanTown Chemical

136280-1KG 136280-250G 136280-5KG
BT136280-1KGEA 115.94 USD
BT136280-1KG BT136280-250G BT136280-5KG
Silica gel desiccant, with indicator
Silica gel

CAS: 63231-67-4; EC No: 231-545-4; MDL No: MFCD00011232 6-16 Mesh Granules Melting Point: 1600°; Boiling Point: 2230° Hygroscopic


  • Hygroscopic
  • 6-16 Mesh Granules

Formula: SiO₂
Boiling Pt: 2230 °C
Melting Pt: >1600 °C
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR