Methyl chloroacetate ≥99%

Supplier: BeanTown Chemical

136245-500G
BT136245-2.5KGEA 110.85 USD
BT136245-2.5KG BT136245-500G
Methyl chloroacetate ≥99%
Methyl chloroacetate

CAS: 96-34-4; EC No: 202-501-1; MDL No: MFCD00000931; RTECS: AF9500000 UN No: UN2295; Haz Class: 6.1 (3); Packing Group: I Liquid; Linear Formula: ClCH2COOCH3; Molecular Formula: C3H5ClO2; MW: 108.52 Melting Point: -33°; Boiling Point: 128-130°; Flash point: 47°C (117°F) Density (g/mL): 1.238; Refractive Index: 1.422


  • RTECS: AF9500000
  • Refractive index: 1.422
  • Liquid

Formula: C₃H₅ClO₂
MW: 108.52 g/mol
Boiling Pt: 131
Melting Pt: -33
Density: 1.238 g/mL
Flash Pt: 47 °C (117 °F)
MDL Number: MFCD00000931
CAS Number: 96-34-4
EINECS: 202-501-1
UN: 2295
ADR: 6.1 I
Merck Index: 12,06122

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR