Hexachloroplatinic(IV) acid hexahydrate ≥99.9%
Supplier: BeanTown Chemical
Synonyms:
Hydrogen hexachloroplatinate (IV) hexahydrate, Chloroplatinic acid hexahydrate, Dihydrogen hexachloroplatinate (IV) hexahydrate
136200-1G
136200-5G
BT136200-1GEA
101.1
USD
BT136200-1G
BT136200-5G
Hexachloroplatinic(IV) acid hexahydrate ≥99.9%
Hexachloroplatinic (IV) acid hexahydrate
CAS: 18497-13-7; EC No: 241-010-7; MDL No: MFCD00149910; RTECS: TP1500000
UN No: UN2507; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: H2PtCl6·6H2O; MW: 517.91
Melting Point: 60° (decomposes)
Hygroscopic
- RTECS: TP1500000
- Hygroscopic
- Crystalline
Formula:
H₂PtCl₆·6H₂O Melting Pt: 60 °C (dec.) |
MDL Number:
MFCD00149910 CAS Number: 18497-13-7 EINECS: 241-010-7 UN: 2507 ADR: 8,III |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...