1-Benzyl-5-phenylbarbituric acid ≥98%

Supplier: BeanTown Chemical

136175-25G 136175-5G
BT136175-25GEA 133 USD
BT136175-25G BT136175-5G
1-Benzyl-5-phenylbarbituric acid ≥98%
1-Benzyl-5-phenylbarbituric acid

CAS: 72846-00-5; EC No: 276-940-2; MDL No: MFCD00082750 Solid; Molecular Formula: C17H14N2O3 ; MW: 294.30


  • Solid

Formula: C₁₇H₁₄N₂O₃
MDL Number: MFCD00082750
CAS Number: 72846-00-5
EINECS: 276-940-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR