Strontium oxide, Technical Grade
Supplier: BeanTown Chemical
136040-1KG
136040-250G
136040-5KG
BT136040-1KGEA
336
USD
BT136040-1KG
BT136040-250G
BT136040-5KG
Strontium oxide, Technical Grade
Strontium oxide
CAS: 1314-11-0; EC No: 215-219-9; MDL No: MFCD00049558
UN No: UN3262; Haz Class: 8; Packing Group: II
Powder; Linear Formula: SrO; MW: 103.62
Melting Point: 2430°; Boiling Point: 3000°
Density (g/mL): 4.7
Moisture Sensitive
- Moisture Sensitive
- Powder
Formula:
SrO Boiling Pt: 3000 °C Melting Pt: 2430 °C Density: 4.7 g/mL |
MDL Number:
MFCD00049558 CAS Number: 1314-11-0 EINECS: 215-219-9 UN: 3262 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...