Strontium oxide, Technical Grade

Supplier: BeanTown Chemical

136040-1KG 136040-250G 136040-5KG
BT136040-1KGEA 336 USD
BT136040-1KG BT136040-250G BT136040-5KG
Strontium oxide, Technical Grade
Strontium oxide

CAS: 1314-11-0; EC No: 215-219-9; MDL No: MFCD00049558 UN No: UN3262; Haz Class: 8; Packing Group: II Powder; Linear Formula: SrO; MW: 103.62 Melting Point: 2430°; Boiling Point: 3000° Density (g/mL): 4.7 Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: SrO
Boiling Pt: 3000 °C
Melting Pt: 2430 °C
Density: 4.7 g/mL
MDL Number: MFCD00049558
CAS Number: 1314-11-0
EINECS: 215-219-9
UN: 3262
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR