Copper(II) sulfide, anhydrous ≥99.98%

Supplier: BeanTown Chemical

Synonyms: Copper sulfide, Cupric sulfide, Copper sulphide, Cupric sulphide, Copper(II)sulfide, Copper(II)sulphide

ampouled under argon

135890-10G 135890-50G
BT135890-10GEA 220 USD
BT135890-10G BT135890-50G
Copper(II) sulfide, anhydrous ≥99.98%
Copper (II) sulphide

CAS: 1317-40-4; EC No: 215-271-2; MDL No: MFCD00016066 Powder; Molecular Formula: CuS ; MW: 95.60 Melting Point: 220° (decomposes) Density (g/mL): 4.6 Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • Powder

Formula: CuS
Melting Pt: 220 °C (dec.)
Density: 4.6 g/mL
MDL Number: MFCD00016066
CAS Number: 1317-40-4
EINECS: 215-271-2

Order Now

Specification Test Results

Assay 99.98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR