Glycine ≥99%
Supplier: BeanTown Chemical
Synonyms:
Aminoacetic acid, Glycocoll, H-Gly-OH
135880-10KG
135880-2.5KG
135880-500G
BT135880-10KGEA
443.85
USD
BT135880-10KG
BT135880-2.5KG
BT135880-500G
Glycine ≥99%
Glycine
CAS: 56-40-6; EC No: 200-272-2; MDL No: MFCD00008131; RTECS: MB7600000
Powder; Linear Formula: NH2CH2CO2H; MW: 75.07
Melting Point: 240° (decomposes)
- RTECS: MB7600000
- Powder
Formula:
NH₂CH₂COOH Melting Pt: 240 °C (dec.) |
MDL Number:
MFCD00008131 CAS Number: 56-40-6 EINECS: 200-272-2 |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...