Diiron arsenide ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Iron arsenide
135845-10G
135845-50G
BT135845-10GEA
400
USD
BT135845-10G
BT135845-50G
Diiron arsenide ≥99.5%
Diiron arsenide
CAS: 12005-88-8; EC No: 234-474-7; MDL No: MFCD00151533
UN No: UN1557; Haz Class: 6.1; Packing Group: II
6mm & Down Lump; Molecular Formula: AsFe2; MW: 186.62
Melting Point:
- 6mm & Down Lump
Formula:
AsFe₂ Melting Pt: >400 °C |
MDL Number:
MFCD00151533 CAS Number: 12005-88-8 EINECS: 234-474-7 UN: 1557 ADR: 6.1,II |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...