Isoamyl alcohol ≥98.5% ACS
Supplier: BeanTown Chemical
CAS: 123-51-3; EC No: 204-633-5; MDL No: MFCD00002934; RTECS: EL5425000
UN No: UN1105; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3)2CHCH2CH2OH; Molecular Formula: C5H12O; MW: 88.15
Melting Point: -117°; Boiling Point: 130°; Flash point: 43°C (109°F)
Density (g/mL): 0.809; Refractive Index: 1.406
- RTECS: EL5425000
- Refractive index: 1.406
- Liquid
Formula:
(CH₃)₂CHCH₂CH₂OH Boiling Pt: 130 °C Melting Pt: –117 °C Density: 0.809 g/mL Flash Pt: 43 °C (109 °F) |
MDL Number:
MFCD00002934 CAS Number: 123-51-3 EINECS: 204-633-5 UN: 1105 ADR: 3,II |
|
Specification Test Results
Assay | 98.5% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...