Ethyl cinnamate ≥98%

Supplier: BeanTown Chemical

Synonyms: NSC 6773, 3-Phenyl-acrylicacidethylester, Ethyl 3-phenyl-2-propenoate, Cinnamic acid ethyl ester, 3-Phenyl-2-propenoic acid ethyl ester

135800-100G 135800-2.5KG 135800-500G
BT135800-100GEA 24 USD
BT135800-100G BT135800-2.5KG BT135800-500G
Ethyl cinnamate ≥98%
Ethyl cinnamate

CAS: 103-36-6; EC No: 203-104-6; MDL No: MFCD00009189; RTECS: GD9010000 Liquid; Linear Formula: C6H5CH=CHCOOCH2CH3; Molecular Formula: C11H12O2; MW: 176.21 Melting Point: 6-8°; Boiling Point: 271°; Flash point: 110°C (230°F) Density (g/mL): 1.049; Refractive Index: 1.558


  • RTECS: GD9010000
  • Refractive index: 1.558
  • Liquid

Formula: C₁₁H₁₂O₂
MW: 176.21 g/mol
Boiling Pt: 271
Melting Pt: 7
Density: 1.049 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00009189
CAS Number: 103-36-6
EINECS: 203-104-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR