Ethyl cinnamate ≥98%
Supplier: BeanTown Chemical
Synonyms:
NSC 6773, 3-Phenyl-acrylicacidethylester, Ethyl 3-phenyl-2-propenoate, Cinnamic acid ethyl ester, 3-Phenyl-2-propenoic acid ethyl ester
135800-100G
135800-2.5KG
135800-500G
BT135800-100GEA
24
USD
BT135800-100G
BT135800-2.5KG
BT135800-500G
Ethyl cinnamate ≥98%
Ethyl cinnamate
CAS: 103-36-6; EC No: 203-104-6; MDL No: MFCD00009189; RTECS: GD9010000
Liquid; Linear Formula: C6H5CH=CHCOOCH2CH3; Molecular Formula: C11H12O2; MW: 176.21
Melting Point: 6-8°; Boiling Point: 271°; Flash point:
- RTECS: GD9010000
- Refractive index: 1.558
- Liquid
Formula:
C₁₁H₁₂O₂ MW: 176.21 g/mol Boiling Pt: 271 Melting Pt: 7 Density: 1.049 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00009189 CAS Number: 103-36-6 EINECS: 203-104-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...