Sulfuric acid 0.02 N in water volumetric solution

Supplier: BeanTown Chemical

135780-1L 135780-20L 135780-4L
BT135780-1LEA 31.91 USD
BT135780-1L BT135780-20L BT135780-4L
Sulfuric acid 0.02 N in water volumetric solution
Sulfuric acid

CAS: 7664-93-9; MDL No: MFCD00064589 Liquid; Linear Formula: H2SO4; MW: 98.08


  • Liquid

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 2796
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR