Rubidium carbonate ≥99.95%
Supplier: BeanTown Chemical
135675-1G
135675-25G
135675-5G
BT135675-1GEA
38.73
USD
BT135675-1G
BT135675-25G
BT135675-5G
Rubidium carbonate ≥99.95%
Rubidium carbonate
CAS: 584-09-8; EC No: 209-530-9; MDL No: MFCD00011188; RTECS: FG0650000
Powder; Linear Formula: Rb2CO3; MW: 230.95
Melting Point: 837°
Hygroscopic
- RTECS: FG0650000
- Hygroscopic
- Powder
Formula:
CO₃Rb₂ Melting Pt: 837 °C |
MDL Number:
MFCD00011188 CAS Number: 584-09-8 EINECS: 209-530-9 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...