Pyridinium p-toluenesulfonate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Pyridin-1-ium 4-methylbenzenesulfonate, Pyridin-1-ium 4-methylbenzenesulphonate
135655-100G
135655-25G
135655-500G
BT135655-100GEA
183
USD
BT135655-100G
BT135655-25G
BT135655-500G
Pyridinium p-toluenesulfonate ≥98%
Pyridinium p-toluenesulphonate
CAS: 24057-28-1; EC No: 246-002-7; MDL No: MFCD00013108
Crystalline/Powder; Molecular Formula: C12H13NO3S; MW: 251.31
Melting Point: 117-119°
Moisture Sensitive
- Moisture Sensitive
- Crystalline/Powder
Formula:
C₁₂H₁₃NO₃S MW: 251.31 g/mol Melting Pt: 117…119 °C |
MDL Number:
MFCD00013108 CAS Number: 24057-28-1 EINECS: 246-002-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...